missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, 98+%

Catalog No. p-7022907
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589030 250 g
AAA158900B 1000 g
AAA158900I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1589030 Supplier Thermo Scientific Chemicals Supplier No. A1589030
Only null left

2-Mercaptoethanol, CAS # 60-24-2, is commonly used as a reducing agent in laboratories and has a variety of applications such as solubilizing proteins. | CAS: 60-24-2 | C2H6OS | 78.13 g/mol

Solubilizes proteins by reducing disulfide linkages2-Mercaptoethanol is used in the preparation of nano-graphene for cellular imaging and drug delivery as well as multifunctional polymeric micelle. It acts as a reducing agent used in electrophoresis, amino acid detection, and distinguishing ssDNA/dsDNA. It is also employed as a standard buffer. It is also used in the preparation of PVC heat stabilizers and as a chain transfer agent in the manufacture of PVC. Further, it is used in some RNA isolation procedures to eliminate ribonuclease. It is utilized as a corrosion inhibitor and ore floatation agent. In biochemistry, it is useful to study the activity of the immune system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 2-Mercaptoethanol is a reducing agent that can be used in studies to solubilize proteins by reducing disulfide linkages

Application

• 2-Mercaptoethanol acts as a reducing agent in electrophoresis, amino acid detection, and distinguishing ssDNA/dsDNA
• It can also be used as a standard buffer
• It can also be used in some RNA isolation procedures to eliminate ribonuclease
• In biochemistry, it is useful for studying the activity of the immune system
• Furthermore, it can be used in laboratory applications involving preparation of multifunctional polymeric micelles and nano-graphene for cellular imaging

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Melting Point -100°C
Density 1.114
Boiling Point 156°C to 158°C
Flash Point 73°C (163°F)
Odor Unpleasant
Refractive Index 1.5
Quantity 250 g
UN Number UN2966
Beilstein 773648
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,5869
Solubility Information Miscible with water,alcohol,ether,benzene and other organic solvents.
IUPAC Name 2-sulfanylethan-1-ol
Formula Weight 78.13
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less
Hazard Category H227-H301+H331-H310-H315-H317-H318-H373
Hazard Statement GHS H Statement
H301-H310-H318-H227-H315-H317-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P311-P314-P330-P333+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THIOGLYCOL
EINECSNumber 200-464-6
RTECSNumber KL5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only