missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptoethanol, ≥98%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219024290
Specifications
| 2-Mercaptoethanol | |
| Colorless to Yellow | |
| ≥98% | |
| MFCD00004890 | |
| 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
| OCCS | |
| 78.13 | |
| CHEBI:41218 | |
| Liquid |
| 60-24-2 | |
| 1.12g/cm3 at 20°C | |
| C2H6OS | |
| 500 mL | |
| DGVVWUTYPXICAM-UHFFFAOYSA-N | |
| 2-sulfanylethan-1-ol | |
| 1567 | |
| 78.13 |
Chemical Identifiers
| 60-24-2 | |
| 78.13 | |
| DGVVWUTYPXICAM-UHFFFAOYSA-N | |
| 1567 | |
| OCCS |
| C2H6OS | |
| MFCD00004890 | |
| 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan | |
| CHEBI:41218 |