missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, ≥98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219024283

 View more versions of this product

Catalog No. ICN19024283


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Mercaptoethanol
Colorless to Yellow
≥98%
MFCD00004890
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
OCCS
78.13
CHEBI:41218
Liquid
60-24-2
1.12g/cm3 at 20°C
C2H6OS
250 mL
DGVVWUTYPXICAM-UHFFFAOYSA-N
2-sulfanylethan-1-ol
1567
78.13

Chemical Identifiers

60-24-2
78.13
DGVVWUTYPXICAM-UHFFFAOYSA-N
1567
OCCS
C2H6OS
MFCD00004890
2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
CHEBI:41218
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions