missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard

CAS: 19767-45-4 | C2H5NaO3S2 | 164.18 g/mol

Supplier:  Thermo Scientific Chemicals 443150100

 View more versions of this product

Catalog No. AC443150100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Mercaptoethanesulfonic acid sodium salt
2% max.
97.5 to 102% min. (On dried substance) (Iodometry)
10 g
mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
XOGTZOOQQBDUSI-UHFFFAOYSA-M
sodium;2-sulfanylethanesulfonate
23662354
98%
19767-45-4
Authentic
C2H5NaO3S2
15,5979
Solubility in water: soluble.
C(CS(=O)(=O)[O-])S.[Na+]
164.18
164.18

Chemical Identifiers

19767-45-4
164.18
mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
sodium;2-sulfanylethanesulfonate
C2H5NaO3S2
XOGTZOOQQBDUSI-UHFFFAOYSA-M
23662354
C(CS(=O)(=O)[O-])S.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 243-285-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only