missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard

Catalog No. p-7077295
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443150050 5 g
AC443150100 10 g
AC443150250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC443150050 Supplier Thermo Scientific Chemicals Supplier No. 443150050
Only null left

CAS: 19767-45-4 | C2H5NaO3S2 | 164.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19767-45-4
Molecular Formula C2H5NaO3S2
Molecular Weight (g/mol) 164.18
InChI Key XOGTZOOQQBDUSI-UHFFFAOYSA-M
Synonym mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
PubChem CID 23662354
IUPAC Name sodium;2-sulfanylethanesulfonate
SMILES C(CS(=O)(=O)[O-])S.[Na+]

Specifications

Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102% min. (On dried substance) (Iodometry)
Quantity 5 g
Merck Index 15,5979
Solubility Information Solubility in water: soluble.
Formula Weight 164.18
Percent Purity 98%
Chemical Name or Material 2-Mercaptoethanesulfonic acid sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 243-285-9

RUO – Research Use Only