missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole 98.0+%, TCI America™

Catalog No. p-7131956
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M005525G 25 g
M0055500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M005525G Supplier TCI America Supplier No. M005525G
Only null left

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.244
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 167.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptobenzothiazole
EINECSNumber (5)-0242
RTECSNumber DL6475000
TSCA Yes