accessibility menu, dialog, popup

UserName

SHNS

CAS RN 149-30-4

IUPAC Name: 2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 2(3H)-benzothiazolethione
Molecular Weight (g/mol): 167.24
Molecular Formula: C7H5NS2
InChi Key: YXIWHUQXZSMYRE-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (6)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (9)
  • (6)
  • (7)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (8)