Learn More
2-Mercaptobenzothiazole, 95%
CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol
Supplier: Thermo Scientific Chemicals 125450010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| 2-Mercaptobenzothiazole | |
| 94.0 | |
| 172.0°C to 182.0°C | |
| 243°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00005781 | |
| 27, II, 233 | |
| 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
| YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
| 3H-1,3-benzothiazole-2-thione | |
| 697993 | |
| 167.24 | |
| Powder |
| 149-30-4 | |
| 100.0 | |
| Yellow | |
| 0.5% max. (60°C, 3 hrs) | |
| 95% | |
| C7H5NS2 | |
| 1 kg | |
| 15, 5935 | |
| Solubility in water: 0.12g/L (25°C). Other solubilities: soluble in alkalies,soluble in alcohol,acetone,benzene and,chloroform | |
| C1=CC=C2C(=C1)NC(=S)S2 | |
| 167.24 | |
| CHEBI:34292 | |
| 95% |
Chemical Identifiers
| 149-30-4 | |
| 167.24 | |
| YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
| 697993 | |
| 3H-1,3-benzothiazole-2-thione |
| C7H5NS2 | |
| MFCD00005781 | |
| 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
| CHEBI:34292 | |
| C1=CC=C2C(=C1)NC(=S)S2 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves.
GHS Signal Word: Warning
EINECSNumber : 205-736-8
RUO – Research Use Only