missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol 98.0+%, TCI America™

Catalog No. M27391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27391G 1 g
M27395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27391G Supplier TCI America Supplier No. M27391G
Only null left

Chemical Identifiers

CAS 368-54-7
Molecular Formula C5H3F3N2OS
Molecular Weight (g/mol) 196.147
MDL Number MFCD00112978
InChI Key YZAOETRYQWFEOY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-trifluoromethyl pyrimidine-2-thiol, 2-mercapto-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl-4-pyrimidinol, 4-hydroxy-2-mercapto-6-trifluoromethyl pyrimidine, 6-trifluoromethyl-2-thiouracil, 2-thioxo-6-trifluoromethyl-2,3-dihydropyrimidin-4 1h-one, 4-hydroxy-2-mercapto-6-trifluoromethylpyrimidine, uracil, 2-thio-6-trifluoromethyl, 2-mercapto-6-trifluoromethyl-pyrimidin-4-ol
PubChem CID 1239796
IUPAC Name 2-sulfanylidene-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES C1=C(NC(=S)NC1=O)C(F)(F)F

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol