missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercapto-6-nitrobenzothiazole 98.0+%, TCI America™
Supplier: TCI America N032625G
Specifications
| 2-Mercapto-6-nitrobenzothiazole | |
| C7H4N2O2S2 | |
| 25 g | |
| QPOZGXKWWKLJDK-UHFFFAOYSA-N | |
| 6-nitro-2,3-dihydro-1,3-benzothiazole-2-thione | |
| 947375 | |
| ≥98.0% (HPLC,T) |
| 4845-58-3 | |
| MFCD00041850 | |
| 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro | |
| [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1 | |
| 212.24 | |
| 212.24 | |
| Crystalline Powder |
Chemical Identifiers
| 4845-58-3 | |
| 212.24 | |
| QPOZGXKWWKLJDK-UHFFFAOYSA-N | |
| 947375 | |
| [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1 |
| C7H4N2O2S2 | |
| MFCD00041850 | |
| 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro | |
| 6-nitro-2,3-dihydro-1,3-benzothiazole-2-thione |
Safety and Handling
RTECSNumber : DL6600000
TSCA : Yes