missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-6-nitrobenzothiazole 98.0+%, TCI America™

Catalog No. p-7133750
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
N03265G 5 g
N032625G 25 g
2 options

Catalog No. N03265G

Supplier: TCI America N03265G

Only null left

Chemical Identifiers

CAS 4845-58-3
Molecular Formula C7H4N2O2S2
Molecular Weight (g/mol) 212.24
MDL Number MFCD00041850
InChI Key QPOZGXKWWKLJDK-UHFFFAOYSA-N
Synonym 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro
PubChem CID 947375
IUPAC Name 6-nitro-2,3-dihydro-1,3-benzothiazole-2-thione
SMILES [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1

Specifications

Quantity 5 g
Formula Weight 212.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole
RTECSNumber DL6600000
TSCA Yes