missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. p-7133144
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22995G 5 g
M229925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22995G Supplier TCI America Supplier No. M22995G

Chemical Identifiers

CAS 76041-72-0
Molecular Formula C6H4F3NS
Molecular Weight (g/mol) 179.16
MDL Number MFCD00128893
InChI Key YFSCXLSUZZXNHW-UHFFFAOYSA-N
Synonym 2-mercapto-5-trifluoromethyl pyridine, 5-trifluoromethyl pyridine-2 1h-thione, 5-trifluoromethyl pyridine-2-thiol, 2-mercapto-5-trifluoromethylpyridine, 2 1h-pyridinethione, 5-trifluoromethyl, 5-trifluoromethyl-2-pyridinethiol, 5-trifluoromethyl-2 1h-pyridinethione, 5-trifluoromethyl-1h-pyridine-2-thione, 5-trifluoromethyl pyridin-2-thiol, 5-trifluoromethyl-2-mercaptopyridine
PubChem CID 2775246
IUPAC Name 5-(trifluoromethyl)-1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1C(F)(F)F

Specifications

Melting Point 150°C
Color Yellow
Quantity 5 g
Formula Weight 179.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-(trifluoromethyl)pyridine
TSCA No
Recommended Storage Refrigerator