accessibility menu, dialog, popup

UserName

CAS RN 76041-72-0

IUPAC Name: 5-(trifluoromethyl)-1,2-dihydropyridine-2-thione
Synonyms: 5-(trifluoromethyl)-1H-pyridine-2-thione
Molecular Weight (g/mol): 179.16
Molecular Formula: C6H4F3NS
InChi Key: YFSCXLSUZZXNHW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNC(=S)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results