missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4-methylpyrimidine hydrochloride, 99%

Catalog No. AAA1314522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1314522 100 g
AAA1314514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1314522 Supplier Thermo Scientific Chemicals Supplier No. A1314522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6959-66-6 | C5H7ClN2S | 162.635 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6959-66-6
Molecular Formula C5H7ClN2S
Molecular Weight (g/mol) 162.635
MDL Number MFCD00012783
InChI Key UQJLPBLXSJWAKG-UHFFFAOYSA-N
Synonym 2-mercapto-4-methylpyrimidine hydrochloride, 4-methylpyrimidine-2-thiol hydrochloride, 4-methylpyrimidine-2 1h-thione hydrochloride, 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride, 4-methylpyrimidine-2-thiol hcl, 2 1h-pyrimidinethione, 4-methyl-, monohydrochloride, c5h6n2s.clh, 2-mercapto-4-methyl pyrimidine hydrochloride, pubchem21990, acmc-1b3ky
PubChem CID 2723697
IUPAC Name 6-methyl-1H-pyrimidine-2-thione;hydrochloride
SMILES CC1=CC=NC(=S)N1.Cl

Specifications

Melting Point ∼265°C (decomposition)
Quantity 100 g
Beilstein 3691520
Formula Weight 162.64
Percent Purity 99%
Chemical Name or Material 2-Mercapto-4-methylpyrimidine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 230-152-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only