accessibility menu, dialog, popup

UserName

CAS RN 6959-66-6

IUPAC Name: 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloridyl
Synonyms: 4-methyl-3H-pyrimidine-2-thione hydrochloridyl
Molecular Weight (g/mol): 161.63
Molecular Formula: C5H6ClN2S
InChi Key: BQUGSWHPLSUHNI-UHFFFAOYSA-N
SMILES: [Cl].CC1=CC=NC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results