missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercapto-4(3H)-quinazolinone 98.0+%, TCI America™
Supplier: TCI America M10895G
Specifications
| 2-Mercapto-4(3H)-quinazolinone | |
| White | |
| MFCD00006885 | |
| 2-Thio-4(3H)-quinazolinone | |
| C1=CC=C2C(=C1)C(=O)NC(=S)N2 | |
| 178.209 | |
| 178.21 | |
| Crystalline Powder |
| 13906-09-7 | |
| C8H6N2OS | |
| 5 g | |
| PUPFOFVEHDNUJU-UHFFFAOYSA-N | |
| 2-sulfanylidene-1H-quinazolin-4-one | |
| 683940 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 13906-09-7 | |
| 178.209 | |
| PUPFOFVEHDNUJU-UHFFFAOYSA-N | |
| 683940 | |
| C1=CC=C2C(=C1)C(=O)NC(=S)N2 |
| C8H6N2OS | |
| MFCD00006885 | |
| 2-Thio-4(3H)-quinazolinone | |
| 2-sulfanylidene-1H-quinazolin-4-one |
Safety and Handling
TSCA : No