Learn More
2-Mercapto-1-methylimidazole, 98%
CAS: 60-56-0 | C4H6N2S | 114.17 g/mol
Supplier: Thermo Scientific Chemicals A1309422
Description
Possesses antioxidant properties
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Mercapto-1-methylimidazole | |
| 144°C to 147°C | |
| Stench | |
| MFCD00179321 | |
| 108646 | |
| methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol | |
| PMRYVIKBURPHAH-UHFFFAOYSA-N | |
| 3-methyl-1H-imidazole-2-thione | |
| 1349907 | |
| 114.17 |
| 60-56-0 | |
| 280°C (decomposition) | |
| C4H6N2S | |
| 100 g | |
| 14,5971 | |
| Freely soluble in water; Soluble in alcohol,chloroform; Sparingly soluble in ether,benzene. | |
| CN1C=CNC1=S | |
| 114.17 | |
| CHEBI:50673 | |
| 98% |
Chemical Identifiers
| 60-56-0 | |
| 114.17 | |
| PMRYVIKBURPHAH-UHFFFAOYSA-N | |
| 1349907 | |
| 3-methyl-1H-imidazole-2-thione |
| C4H6N2S | |
| MFCD00179321 | |
| methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol | |
| CHEBI:50673 | |
| CN1C=CNC1=S |
Safety and Handling
GHS H Statement
H361-H317
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H361fd
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 200-482-4
RTECSNumber : NI8615000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only