accessibility menu, dialog, popup

UserName

CH3NHNS

CAS RN 60-56-0

IUPAC Name: 1-methyl-2,3-dihydro-1H-imidazole-2-thione
Synonyms: methimazole|tapazole|mercazole|thiamazole|basolan|methamazole|methimazol|thiamazol|methiamazole|danantizol
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: PMRYVIKBURPHAH-UHFFFAOYSA-N
SMILES: CN1C=CNC1=S
Percent Purity 
  • (1)
  • (2)
  • (4)
  • (2)
Grade 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)