missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-1-methylimidazole, 98%

Catalog No. AAA1309414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1309414 25 g
AAA1309422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1309414 Supplier Thermo Scientific Chemicals Supplier No. A1309414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60-56-0 | C4H6N2S | 114.17 g/mol

Possesses antioxidant properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00179321
InChI Key PMRYVIKBURPHAH-UHFFFAOYSA-N
Synonym methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol
PubChem CID 1349907
ChEBI CHEBI:50673
IUPAC Name 3-methyl-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Melting Point 144°C to 147°C
Boiling Point 280°C (decomposition)
Odor Stench
Quantity 25 g
Beilstein 108646
Merck Index 14,5971
Solubility Information Freely soluble in water; Soluble in alcohol,chloroform; Sparingly soluble in ether,benzene.
Formula Weight 114.17
Percent Purity 98%
Chemical Name or Material 2-Mercapto-1-methylimidazole
Hazard Category H317-H361fd
Hazard Statement GHS H Statement
H361-H317
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 200-482-4
RTECSNumber NI8615000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only