missing translation for 'onlineSavingsMsg'
Learn More

2-(m-Tolyl)ethanol 98.0+%, TCI America™

Catalog No. p-7132694
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13765G 5 g
M137625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13765G Supplier TCI America Supplier No. M13765G

Chemical Identifiers

CAS 1875-89-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002896
InChI Key KWHVBVJDKLSOTB-UHFFFAOYSA-N
Synonym 3-Methylphenethyl Alcohol, 2-(3-Methylphenyl)ethyl Alcohol, 2-(3-Methylphenyl)ethanol
PubChem CID 74648
IUPAC Name 2-(3-methylphenyl)ethanol
SMILES CC1=CC(=CC=C1)CCO

Specifications

Color Yellow
Boiling Point 243°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(m-Tolyl)ethanol