CAS RN 1875-89-4
CAS RN 1875-89-4
IUPAC Name:
2-(3-methylphenyl)ethan-1-ol
Synonyms:
2-M-cresylethanol
Molecular Weight (g/mol):
136.19
Molecular Formula:
C9H12O
InChi Key:
KWHVBVJDKLSOTB-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(CCO)=C1
1
–
1
of
1
results
1
2-(m-Tolyl)ethanol 98.0+%, TCI America™
CAS: 1875-89-4 Molecular Formula: C9H12O Molecular Weight (g/mol): 136.194 MDL Number: MFCD00002896 InChI Key: KWHVBVJDKLSOTB-UHFFFAOYSA-N Synonym: 3-Methylphenethyl Alcohol, 2-(3-Methylphenyl)ethyl Alcohol, 2-(3-Methylphenyl)ethanol PubChem CID: 74648 IUPAC Name: 2-(3-methylphenyl)ethanol SMILES: CC1=CC(=CC=C1)CCO