missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isocyanate, 97%

Catalog No. AAL1061814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1061814 25 g
AAL1061806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1061814 Supplier Thermo Scientific Chemicals Supplier No. L1061814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56309-56-9 | C10H11NO | 161.204 g/mol

2-Isopropylphenyl isocyanate is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylphenyl isocyanate is used as intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.

Chemical Identifiers

CAS 56309-56-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00013850
InChI Key MWBSQVPLYJZPTG-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
PubChem CID 91797
IUPAC Name 1-isocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=O

Specifications

Density 1.019
Boiling Point 66°C to 67°C (5 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.523
Quantity 25 g
UN Number UN2206
Beilstein 3588928
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylphenyl isocyanate
Show More Show Less
Hazard Category H227-H302+H312-H315-H317-H319-H331-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 260-105-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only