missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenol 98.0+%, TCI America™

Catalog No. i017325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
I017325G 25 g
I0173500G 500 g
2 options

Catalog No. I017325G

Supplier: TCI America I017325G

Only null left

Chemical Identifiers

CAS 88-69-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002224
InChI Key CRBJBYGJVIBWIY-UHFFFAOYSA-N
Synonym 2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
PubChem CID 6943
ChEBI CHEBI:38506
IUPAC Name 2-propan-2-ylphenol
SMILES CC(C)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 g
UN Number 3145
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropylphenol
EINECSNumber (3)-0503&(4)-0057
RTECSNumber SL5900000
TSCA Yes