missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropylimidazole 98.0+%, TCI America™
$41.02 - $290.79
Chemical Identifiers
| CAS | 36947-68-9 |
|---|---|
| Molecular Formula | C6H10N2 |
| Molecular Weight (g/mol) | 110.16 |
| MDL Number | MFCD00014486 |
| InChI Key | FUOZJYASZOSONT-UHFFFAOYSA-N |
| Synonym | 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole |
| PubChem CID | 123457 |
| IUPAC Name | 2-(propan-2-yl)-1H-imidazole |
| SMILES | CC(C)C1=NC=CN1 |
Chemical Identifiers
| 36947-68-9 | |
| 110.16 | |
| FUOZJYASZOSONT-UHFFFAOYSA-N | |
| 123457 | |
| CC(C)C1=NC=CN1 |
| C6H10N2 | |
| MFCD00014486 | |
| 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole | |
| 2-(propan-2-yl)-1H-imidazole |
Specifications
| 36947-68-9 | |
| White-Yellow | |
| MFCD00014486 | |
| 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole | |
| CC(C)C1=NC=CN1 | |
| 110.16 | |
| 110.16 | |
| Crystalline Powder |
| 132°C | |
| C6H10N2 | |
| 25 g | |
| FUOZJYASZOSONT-UHFFFAOYSA-N | |
| 2-(propan-2-yl)-1H-imidazole | |
| 123457 | |
| ≥98.0% (GC,T) | |
| 2-Isopropylimidazole |
Safety and Handling
TSCA : Yes