accessibility menu, dialog, popup

UserName

H3CCH3NHN

CAS RN 36947-68-9

IUPAC Name: 2-(propan-2-yl)-1H-imidazole
Synonyms: 2-isopropyl-1H-imidazole
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: FUOZJYASZOSONT-UHFFFAOYSA-N
SMILES: CC(C)C1=NC=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)