missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylimidazole 98.0+%, TCI America™

Catalog No. I031325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I031325G 25 g
I0313500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I031325G Supplier TCI America Supplier No. I031325G

Chemical Identifiers

CAS 36947-68-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00014486
InChI Key FUOZJYASZOSONT-UHFFFAOYSA-N
Synonym 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
PubChem CID 123457
IUPAC Name 2-(propan-2-yl)-1H-imidazole
SMILES CC(C)C1=NC=CN1

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 g
Formula Weight 110.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropylimidazole