missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylimidazole, 98%

CAS: 36947-68-9 | C6H10N2 | 110.16 g/mol

$72.90 - $229.49

Chemical Identifiers

CAS 36947-68-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00014486
InChI Key FUOZJYASZOSONT-UHFFFAOYSA-N
Synonym 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
PubChem CID 123457
IUPAC Name 2-propan-2-yl-1H-imidazole
SMILES CC(C)C1=NC=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1330722
View Documents
Thermo Scientific Chemicals
A1330722
100 g
Each for $72.90
Only null left
 
AAA1330736
View Documents
Thermo Scientific Chemicals
A1330736
500 g
Each for $229.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

36947-68-9
110.16
FUOZJYASZOSONT-UHFFFAOYSA-N
123457
CC(C)C1=NC=CN1
C6H10N2
MFCD00014486
2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
2-propan-2-yl-1H-imidazole

Specifications

36947-68-9
256°C to 260°C
MFCD00014486
107804
2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
CC(C)C1=NC=CN1
110.16
110.16
2-Isopropylimidazole
126°C to 132°C
C6H10N2
100 g
Hygroscopic
FUOZJYASZOSONT-UHFFFAOYSA-N
2-propan-2-yl-1H-imidazole
123457
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 253-286-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only