missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylimidazole, 98%

Catalog No. AAA1330722
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1330722 100 g
AAA1330736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1330722 Supplier Thermo Scientific Chemicals Supplier No. A1330722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 36947-68-9 | C6H10N2 | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36947-68-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00014486
InChI Key FUOZJYASZOSONT-UHFFFAOYSA-N
Synonym 2-isopropylimidazole, 2-isopropyl-1h-imidazole, 1h-imidazole, 2-1-methylethyl, 2-propan-2-yl-1h-imidazole, unii-a92tuy97g2, 2-isopropylimdazole, 2-isopropylimidazol, 2-iso-propylimidazole, 2-isopropyl imidazole, 2-isopropyl-imidazole
PubChem CID 123457
IUPAC Name 2-propan-2-yl-1H-imidazole
SMILES CC(C)C1=NC=CN1

Specifications

Melting Point 126°C to 132°C
Boiling Point 256°C to 260°C
Quantity 100 g
Beilstein 107804
Sensitivity Hygroscopic
Formula Weight 110.16
Percent Purity 98%
Chemical Name or Material 2-Isopropylimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-286-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only