missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylbenzeneboronic acid, 97%

Catalog No. AAL2011003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2011003 1 g
AAL2011006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2011003 Supplier Thermo Scientific Chemicals Supplier No. L2011003

CAS: 89787-12-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89787-12-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD03411937
InChI Key KTZUVUWIBZMHMC-UHFFFAOYSA-N
Synonym 2-isopropylphenylboronic acid, 2-isopropylphenyl boronic acid, 2-isopropylbenzeneboronic acid, 2-cumylboronic acid, 2-propan-2-ylphenyl boronic acid, o-isopropylphenylboronic acid, 2-isopropylphenboronic acid, 2-isopropylphenyboronic acid, 2-propan-2-yl phenyl boronic acid, boronic acid, 2-1-methylethyl phenyl
PubChem CID 2773477
IUPAC Name (2-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC=C1B(O)O

Specifications

Melting Point 85°C to 86°C
Quantity 1 g
Beilstein 3090298
Formula Weight 164.01
Percent Purity 97%
Chemical Name or Material 2-Isopropylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only