Learn More
2-Isopropylaniline, 97%
CAS: 643-28-7 | C9H13N | 135.21 g/mol
Supplier: Thermo Scientific Chemicals A1467122
Description
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
Specifications
| 2-Isopropylaniline | |
| 0.966 | |
| 95°C (203°F) | |
| 1.5485 | |
| 100 g | |
| Light sensitive | |
| Insoluble in water. | |
| CC(C)C1=CC=CC=C1N | |
| 135.21 | |
| 135.21 |
| 643-28-7 | |
| 214°C to 216°C | |
| C9H13N | |
| MFCD00007720 | |
| 636283 | |
| 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline | |
| YKOLZVXSPGIIBJ-UHFFFAOYSA-N | |
| 2-propan-2-ylaniline | |
| 12561 | |
| 97% |
Chemical Identifiers
| 643-28-7 | |
| 135.21 | |
| YKOLZVXSPGIIBJ-UHFFFAOYSA-N | |
| 12561 |
| C9H13N | |
| MFCD00007720 | |
| 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline | |
| CC(C)C1=CC=CC=C1N |
Safety and Handling
GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c
H302+H312+H332-H336-H373
EINECSNumber : 211-397-7
RTECSNumber : BY4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only