missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylaniline, 97%

Catalog No. AAA1467136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467136 500 g
AAA1467114 25 g
AAA1467122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1467136 Supplier Thermo Scientific Chemicals Supplier No. A1467136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 643-28-7 | C9H13N | 135.21 g/mol

2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive

Chemical Identifiers

CAS 643-28-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007720
InChI Key YKOLZVXSPGIIBJ-UHFFFAOYSA-N
Synonym 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline
PubChem CID 12561
SMILES CC(C)C1=CC=CC=C1N

Specifications

Density 0.966
Boiling Point 214°C to 216°C
Flash Point 95°C (203°F)
Refractive Index 1.5485
Quantity 500 g
Beilstein 636283
Sensitivity Light sensitive
Solubility Information Insoluble in water.
IUPAC Name 2-propan-2-ylaniline
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylaniline
Show More Show Less
Hazard Category H302+H312+H332-H336-H373
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c
EINECSNumber 211-397-7
RTECSNumber BY4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only