missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Isopropylamino)ethanol 99.0+%, TCI America™
$98.79 - $784.76
Chemical Identifiers
| CAS | 109-56-8 |
|---|---|
| Molecular Formula | C5H13NO |
| Molecular Weight (g/mol) | 103.165 |
| MDL Number | MFCD00041755 |
| InChI Key | RILLZYSZSDGYGV-UHFFFAOYSA-N |
| Synonym | 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine |
| PubChem CID | 7994 |
| IUPAC Name | 2-(propan-2-ylamino)ethanol |
| SMILES | CC(C)NCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I022725ML
|
TCI America
I022725ML |
25 mL |
Each for $98.79
|
|
|||||
|
I0227100ML
|
TCI America
I0227100ML |
100 mL |
Each for $257.24
|
|
|||||
|
I0227500ML
|
TCI America
I0227500ML |
500 mL |
Each for $784.76
|
|
|||||
Chemical Identifiers
| 109-56-8 | |
| 103.165 | |
| RILLZYSZSDGYGV-UHFFFAOYSA-N | |
| 7994 | |
| CC(C)NCCO |
| C5H13NO | |
| MFCD00041755 | |
| 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine | |
| 2-(propan-2-ylamino)ethanol |
Specifications
| 109-56-8 | |
| 171°C | |
| MFCD00041755 | |
| 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine | |
| CC(C)NCCO | |
| 103.165 | |
| 103.17 | |
| Liquid |
| Yellow | |
| C5H13NO | |
| 25 mL | |
| RILLZYSZSDGYGV-UHFFFAOYSA-N | |
| 2-(propan-2-ylamino)ethanol | |
| 7994 | |
| ≥99.0% (GC,T) | |
| 2-(Isopropylamino)ethanol |
Safety and Handling
EINECSNumber : (2)-0295
RTECSNumber : KL5080000
TSCA : Yes