missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Isopropylamino)ethanol 99.0+%, TCI America™
Supplier: TCI America I0227100ML
Specifications
| 2-(Isopropylamino)ethanol | |
| Yellow | |
| C5H13NO | |
| 100 mL | |
| RILLZYSZSDGYGV-UHFFFAOYSA-N | |
| 2-(propan-2-ylamino)ethanol | |
| 7994 | |
| ≥99.0% (GC,T) |
| 109-56-8 | |
| 171°C | |
| MFCD00041755 | |
| 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine | |
| CC(C)NCCO | |
| 103.165 | |
| 103.17 | |
| Liquid |
Chemical Identifiers
| 109-56-8 | |
| 103.165 | |
| RILLZYSZSDGYGV-UHFFFAOYSA-N | |
| 7994 | |
| CC(C)NCCO |
| C5H13NO | |
| MFCD00041755 | |
| 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine | |
| 2-(propan-2-ylamino)ethanol |
Safety and Handling
EINECSNumber : (2)-0295
RTECSNumber : KL5080000
TSCA : Yes