missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropyl-d7-thioxanthen-9-one, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| CAS | 1173019-24-3 |
| Physical Form | Neat |
| Linear Formula | C16D7H7 OS |
| MDL Number | MFCD09842833 |
| Synonym | ITX-d7 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 261.39 |
| Grade | Analytical Standard |