missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropyl-5-methylanisole 96.0+%, TCI America™
$87.34 - $252.51
Chemical Identifiers
| CAS | 1076-56-8 |
|---|---|
| Molecular Formula | C11H16O |
| Molecular Weight (g/mol) | 164.248 |
| MDL Number | MFCD01674973 |
| InChI Key | LSQXNMXDFRRDSJ-UHFFFAOYSA-N |
| Synonym | 1-Isopropyl-2-methoxy-4-methylbenzene, 4-Isopropyl-3-methoxytoluene |
| PubChem CID | 14104 |
| IUPAC Name | 2-methoxy-4-methyl-1-propan-2-ylbenzene |
| SMILES | CC1=CC(=C(C=C1)C(C)C)OC |
Chemical Identifiers
| C11H16O | |
| MFCD01674973 | |
| 1-Isopropyl-2-methoxy-4-methylbenzene, 4-Isopropyl-3-methoxytoluene | |
| 2-methoxy-4-methyl-1-propan-2-ylbenzene |
Specifications
| 1076-56-8 | |
| 216°C | |
| MFCD01674973 | |
| 1-Isopropyl-2-methoxy-4-methylbenzene, 4-Isopropyl-3-methoxytoluene | |
| CC1=CC(=C(C=C1)C(C)C)OC | |
| 164.248 | |
| 164.25 | |
| Liquid |
| Yellow | |
| C11H16O | |
| 1993 | |
| LSQXNMXDFRRDSJ-UHFFFAOYSA-N | |
| 2-methoxy-4-methyl-1-propan-2-ylbenzene | |
| 14104 | |
| ≥96.0% (GC) | |
| 2-Isopropyl-5-methylanisole |
Safety and Handling
RTECSNumber : BZ8770000
TSCA : Yes