missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropyl-5-methylanisole 96.0+%, TCI America™
Supplier: TCI America I099625G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C11H16O | |
| MFCD01674973 | |
| 1-Isopropyl-2-methoxy-4-methylbenzene, 4-Isopropyl-3-methoxytoluene | |
| 2-methoxy-4-methyl-1-propan-2-ylbenzene |
Specifications
| 2-Isopropyl-5-methylanisole | |
| Yellow | |
| C11H16O | |
| MFCD01674973 | |
| 1-Isopropyl-2-methoxy-4-methylbenzene, 4-Isopropyl-3-methoxytoluene | |
| CC1=CC(=C(C=C1)C(C)C)OC | |
| 164.248 | |
| 164.25 | |
| Liquid |
| 1076-56-8 | |
| 216°C | |
| 25 g | |
| 1993 | |
| LSQXNMXDFRRDSJ-UHFFFAOYSA-N | |
| 2-methoxy-4-methyl-1-propan-2-ylbenzene | |
| 14104 | |
| ≥96.0% (GC) |
Safety and Handling
RTECSNumber : BZ8770000
TSCA : Yes