missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropyl-2-methacryloyloxyadamantane 98.0+%, TCI America™
$115.57 - $579.71
Chemical Identifiers
| CAS | 297156-50-4 |
|---|---|
| Molecular Formula | C17H26O2 |
| Molecular Weight (g/mol) | 262.393 |
| MDL Number | MFCD22571643 |
| InChI Key | ZMAOPHHNBQIJOQ-UHFFFAOYSA-N |
| Synonym | 2-Isopropyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Isopropyl-2-adamantyl Ester |
| PubChem CID | 12097425 |
| IUPAC Name | (2-propan-2-yl-2-adamantyl) 2-methylprop-2-enoate |
| SMILES | CC(C)C1(C2CC3CC(C2)CC1C3)OC(=O)C(=C)C |
Chemical Identifiers
| 297156-50-4 | |
| 262.393 | |
| ZMAOPHHNBQIJOQ-UHFFFAOYSA-N | |
| 12097425 | |
| CC(C)C1(C2CC3CC(C2)CC1C3)OC(=O)C(=C)C |
| C17H26O2 | |
| MFCD22571643 | |
| 2-Isopropyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Isopropyl-2-adamantyl Ester | |
| (2-propan-2-yl-2-adamantyl) 2-methylprop-2-enoate |
Specifications
| 297156-50-4 | |
| White | |
| MFCD22571643 | |
| 2-Isopropyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Isopropyl-2-adamantyl Ester | |
| CC(C)C1(C2CC3CC(C2)CC1C3)OC(=O)C(=C)C | |
| 262.393 | |
| 262.39 | |
| Crystalline Powder |
| 40°C | |
| C17H26O2 | |
| 5 g | |
| ZMAOPHHNBQIJOQ-UHFFFAOYSA-N | |
| (2-propan-2-yl-2-adamantyl) 2-methylprop-2-enoate | |
| 12097425 | |
| ≥98.0% (GC) | |
| 2-Isopropyl-2-methacryloyloxyadamantane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator