accessibility menu, dialog, popup

UserName

CAS RN 297156-50-4

H3CH3COOCH2H3C

CAS RN 297156-50-4

IUPAC Name: 2-(propan-2-yl)adamantan-2-yl 2-methylprop-2-enoate
Synonyms: 2-isopropyladamantan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 262.39
Molecular Formula: C17H26O2
InChi Key: ZMAOPHHNBQIJOQ-UHFFFAOYSA-N
SMILES: CC(C)C1(OC(=O)C(C)=C)C2CC3CC(C2)CC1C3
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results