missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenol, 97%

Catalog No. AAL0736006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0736006 5 g
AAL0736014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0736006 Supplier Thermo Scientific Chemicals Supplier No. L0736006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4812-20-8 | C9H12O2 | 152.193 g/mol

2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4812-20-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002184
InChI Key ZNCUUYCDKVNVJH-UHFFFAOYSA-N
Synonym 2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a
PubChem CID 20949
ChEBI CHEBI:38547
IUPAC Name 2-propan-2-yloxyphenol
SMILES CC(C)OC1=CC=CC=C1O

Specifications

Density 1.032
Boiling Point 100°C to 102°C (11 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.515
Quantity 5 g
Beilstein 1937063
Solubility Information Soluble in water.
Formula Weight 152.19
Percent Purity 97%
Chemical Name or Material 2-Isopropoxyphenol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 225-379-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only