missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethanol, 99%

CAS: 109-59-1 | C5H12O2 | 104.15 g/mol

$47.30 - $165.22

Chemical Identifiers

CAS 109-59-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002866
InChI Key HCGFUIQPSOCUHI-UHFFFAOYSA-N
Synonym 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
PubChem CID 7996
IUPAC Name 2-propan-2-yloxyethanol
SMILES CC(C)OCCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL13241AE
View Documents
Thermo Scientific Chemicals
L13241AE
100 mL
Each for $47.30
Only null left
 
AAL13241AU
View Documents
Thermo Scientific Chemicals
L13241AU
1000 mL
Each for $87.55
Only null left
 
AAL132410F
View Documents
Thermo Scientific Chemicals
L132410F
2500 mL
Each for $165.22
Only null left
 
Description

Description

2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

109-59-1
104.15
HCGFUIQPSOCUHI-UHFFFAOYSA-N
7996
CC(C)OCCO
C5H12O2
MFCD00002866
2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
2-propan-2-yloxyethanol

Specifications

109-59-1
0.903
45°C (113°F)
1.41
MFCD00002866
UN1992
2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
HCGFUIQPSOCUHI-UHFFFAOYSA-N
2-propan-2-yloxyethanol
7996
99%
-60°C
143°C
C5H12O2
(CH3)2CHO(CH2)2OH
100 mL
1732184
It is soluble in water.
CC(C)OCCO
104.15
104.15
2-Isopropoxyethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H319
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P363-P370+P378q-P501c

H226-H312+H332-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 203-685-6

RTECSNumber : KL5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only