missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethanol, 99%

CAS: 109-59-1 | C5H12O2 | 104.15 g/mol

Supplier:  Thermo Scientific Chemicals L132410F

 View more versions of this product

Catalog No. AAL132410F


Only null left
Explore available promotions

Description

Description

2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Isopropoxyethanol
-60°C
143°C
C5H12O2
(CH3)2CHO(CH2)2OH
2500 mL
1732184
It is soluble in water.
CC(C)OCCO
104.15
104.15
109-59-1
0.903
45°C (113°F)
1.41
MFCD00002866
UN1992
2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
HCGFUIQPSOCUHI-UHFFFAOYSA-N
2-propan-2-yloxyethanol
7996
99%

Chemical Identifiers

109-59-1
104.15
HCGFUIQPSOCUHI-UHFFFAOYSA-N
7996
CC(C)OCCO
C5H12O2
MFCD00002866
2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
2-propan-2-yloxyethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H319
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P363-P370+P378q-P501c

H226-H312+H332-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 203-685-6

RTECSNumber : KL5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only