missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropenylnaphthalene 98.0+%, TCI America™
$126.76 - $365.35
Chemical Identifiers
| CAS | 3710-23-4 |
|---|---|
| Molecular Formula | C13H12 |
| Molecular Weight (g/mol) | 168.24 |
| MDL Number | MFCD00191524 |
| InChI Key | ANCUXNXTHQXICN-UHFFFAOYSA-N |
| PubChem CID | 77301 |
| IUPAC Name | 2-(prop-1-en-2-yl)naphthalene |
| SMILES | CC(=C)C1=CC=C2C=CC=CC2=C1 |
Chemical Identifiers
| 3710-23-4 | |
| 168.24 | |
| ANCUXNXTHQXICN-UHFFFAOYSA-N | |
| 2-(prop-1-en-2-yl)naphthalene |
| C13H12 | |
| MFCD00191524 | |
| 77301 | |
| CC(=C)C1=CC=C2C=CC=CC2=C1 |
Specifications
| 3710-23-4 | |
| White-Yellow | |
| C13H12 | |
| 5 g | |
| CC(=C)C1=CC=C2C=CC=CC2=C1 | |
| 168.24 | |
| 168.24 | |
| Crystalline Powder |
| 55°C | |
| 155°C | |
| MFCD00191524 | |
| ANCUXNXTHQXICN-UHFFFAOYSA-N | |
| 2-(prop-1-en-2-yl)naphthalene | |
| 77301 | |
| ≥98.0% (GC) | |
| 2-Isopropenylnaphthalene |
Safety and Handling
EINECSNumber : (4)-0313
TSCA : No
Recommended Storage : Refrigerator