accessibility menu, dialog, popup

UserName

CH3H2C

CAS RN 3710-23-4

IUPAC Name: 2-(prop-1-en-2-yl)naphthalene
Synonyms: 2-(prop-1-en-2-yl)naphthalene
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: ANCUXNXTHQXICN-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results