missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropenylnaphthalene 98.0+%, TCI America™

Catalog No. I04135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04135G 5 g
I041325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04135G Supplier TCI America Supplier No. I04135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3710-23-4
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00191524
InChI Key ANCUXNXTHQXICN-UHFFFAOYSA-N
PubChem CID 77301
IUPAC Name 2-(prop-1-en-2-yl)naphthalene
SMILES CC(=C)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropenylnaphthalene
EINECSNumber (4)-0313
TSCA No
Recommended Storage Refrigerator