missing translation for 'onlineSavingsMsg'
Learn More

2-Isonitrosopropiophenone 98.0+%, TCI America™

Catalog No. P021125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P021125G Supplier TCI America Supplier No. P021125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-51-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00002115
InChI Key YPINLRNGSGGJJT-JXMROGBWSA-N
Synonym 1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone
PubChem CID 9566063
IUPAC Name (2E)-2-(N-hydroxyimino)-1-phenylpropan-1-one
SMILES C\C(=N/O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isonitrosopropiophenone
RTECSNumber UH2975000
TSCA Yes