missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodotoluene (stabilized with Copper chip) 98.0+%, TCI America™
Supplier: TCI America I021725G
Specifications
| 2-Iodotoluene (stabilized with Copper chip) | |
| 211°C | |
| MFCD00001042 | |
| 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07 | |
| CC1=CC=CC=C1I | |
| 218.037 | |
| CHEBI:39813 | |
| ≥98.0% (GC) |
| 615-37-2 | |
| C7H7I | |
| 25 g | |
| RINOYHWVBUKAQE-UHFFFAOYSA-N | |
| 1-iodo-2-methylbenzene | |
| 5128 | |
| 218.04 | |
| Liquid |
Chemical Identifiers
| 615-37-2 | |
| 218.037 | |
| RINOYHWVBUKAQE-UHFFFAOYSA-N | |
| 5128 | |
| 1-iodo-2-methylbenzene |
| C7H7I | |
| MFCD00001042 | |
| 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07 | |
| CHEBI:39813 | |
| CC1=CC=CC=C1I |
Safety and Handling
TSCA : Yes