missing translation for 'onlineSavingsMsg'
Learn More

2-Iodothiophene 98.0+%, TCI America™

Catalog No. p-7131369
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I037425G 25 g
I0374100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I037425G Supplier TCI America Supplier No. I037425G
Only null left

Chemical Identifiers

CAS 3437-95-4
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.032
MDL Number MFCD00005424
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
PubChem CID 18921
IUPAC Name 2-iodothiophene
SMILES C1=CSC(=C1)I

Specifications

Melting Point -40°C
Color Green-Yellow
Boiling Point 182°C
Quantity 25 g
Formula Weight 210.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodothiophene