Learn More
2-Iodothiophene, 98%, stab. with copper
CAS: 3437-95-4 | C4H3IS | 210.032 g/mol
Supplier: Thermo Scientific Chemicals A1289822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Iodothiophene | |
| 3437-95-4 | |
| 2.064 | |
| 71°C (159°F) | |
| 1.652 | |
| 100 g | |
| Light sensitive | |
| ROIMNSWDOJCBFR-UHFFFAOYSA-N | |
| 2-iodothiophene | |
| 18921 | |
| 98% |
| stabilized with copper | |
| -40°C | |
| 182°C | |
| C4H3IS | |
| MFCD00005424 | |
| 104666 | |
| thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene | |
| C1=CSC(=C1)I | |
| 210.032 | |
| 210.04 |
Chemical Identifiers
| 3437-95-4 | |
| 210.032 | |
| ROIMNSWDOJCBFR-UHFFFAOYSA-N | |
| 18921 | |
| C1=CSC(=C1)I |
| C4H3IS | |
| MFCD00005424 | |
| thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene | |
| 2-iodothiophene |
Safety and Handling
GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H317-H318-H335
EINECSNumber : 222-342-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only