missing translation for 'onlineSavingsMsg'
Learn More

2-Iodothiophene, 98%, stab. with copper

Catalog No. AAA1289814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1289814 25 g
AAA1289822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1289814 Supplier Thermo Scientific Chemicals Supplier No. A1289814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3437-95-4 | C4H3IS | 210.032 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3437-95-4
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.032
MDL Number MFCD00005424
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
PubChem CID 18921
IUPAC Name 2-iodothiophene
SMILES C1=CSC(=C1)I

Specifications

Melting Point -40°C
Density 2.064
Boiling Point 182°C
Flash Point 71°C (159°F)
Refractive Index 1.652
Quantity 25 g
Beilstein 104666
Sensitivity Light sensitive
Formula Weight 210.04
Percent Purity 98%
Chemical Name or Material 2-Iodothiophene, stabilized with copper
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
EINECSNumber 222-342-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only