missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoresorcinol 97.0+%, TCI America™

Catalog No. I07881G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07881G Supplier TCI America Supplier No. I07881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41046-67-7
Molecular Formula C6H5IO2
Molecular Weight (g/mol) 236.008
MDL Number MFCD09701440
InChI Key BNJXHRMYHDWZKL-UHFFFAOYSA-N
Synonym 2-iodoresorcinol, 2-iodo-1,3-benzenediol, 1,3-benzenediol, 2-iodo, 2-iodo-1,3-dihydroxybenzene, iodoresorcinol, ambkkkkk491, acmc-1aol8, 1,3-benzenediol,2-iodo, 2-iodo-benzene-1,3-diol, 2-iodo-1,3-benzenediol #
PubChem CID 616833
IUPAC Name 2-iodobenzene-1,3-diol
SMILES C1=CC(=C(C(=C1)O)I)O

Specifications

Melting Point 108°C
Quantity 1 g
Formula Weight 236.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodoresorcinol
TSCA No
Recommended Storage Freezer